3'-Ethoxy-2',4'-difluoroacetophenone 97%, JRD

Warning

H31869-03 H31869-06
AAH31869-06EA 218.67 USD
AAH31869-06 AAH31869-03
3'-Ethoxy-2',4'-difluoroacetophenone 97%, JRD
3'-Ethoxy-2',4'-difluoroacetophenone
Formula: C₁₀H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09258734
CAS Number: 1017778-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR