Tris[2-(dimethylamino)ethyl]amine ≥99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Me6TREN
H31853-06
H31853-14
AAH31853-14EA
699.3
USD
AAH31853-14
AAH31853-06
Tris[2-(dimethylamino)ethyl]amine ≥99%
Tris[2-(dimethylamino)ethyl]amine
Formula:
C₁₂H₃₀N₄ MW: 230.40 g/mol Boiling Pt: 70…71 °C/0.5 mm Storage Temperature: Ambient |
MDL Number:
MFCD00015607 CAS Number: 33527-91-2 UN: 2735 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...