5-Bromo-3-(trifluoromethoxy)salicylaldehyde 99%

Warning

Synonyms: 5-Bromo-2-hydroxy-3-(trifluoromethoxy)benzaldehyde

H31842-MD H31842-03
AAH31842-MDEA 81.98 USD
AAH31842-MD AAH31842-03
5-Bromo-3-(trifluoromethoxy)salicylaldehyde 99%
5-Bromo-3-(trifluoromethoxy)salicylaldehyde
Formula: C₈H₄BrF₃O₃
MW: 285.02
Melting Pt: 44…48 °C
Storage Temperature: Ambient
MDL Number: MFCD03788532
CAS Number: 497959-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR