3-Methyl-5-(trifluoromethoxy)benzylamine 97%, JRD
|
Danger
|
H31840-MD
AAH31840-MDEA
72.38
USD
AAH31840-MD
3-Methyl-5-(trifluoromethoxy)benzylamine 97%, JRD
3-Methyl-5-(trifluoromethoxy)benzylamine
Formula:
C₉H₁₀F₃NO Storage Temperature: Ambient |
MDL Number:
MFCD09025379 CAS Number: 916420-60-5 UN: 2735 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...