8-Fluoro-4-quinolinol 97%

Danger

Synonyms: 8-Fluoroquinolin-4-ol8-Fluoro-4-hydroxyquinoline

H31831-03
AAH31831-03EA 79.74 USD
AAH31831-03
8-Fluoro-4-quinolinol 97%
8-Fluoro-4-quinolinol
Formula: C₉H₆FNO
MW: 163.15 g/mol
Melting Pt: 280…282 °C
Storage Temperature: Ambient
MDL Number: MFCD00272395
CAS Number: 63010-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR