3-Bromo-5-(trifluoromethoxy)benzyl alcohol 97%, JRD

Warning

H31815-03 H31815-06
AAH31815-06EA 297.48 USD
AAH31815-06 AAH31815-03
3-Bromo-5-(trifluoromethoxy)benzyl alcohol 97%, JRD
3-Bromo-5-(trifluoromethoxy)benzyl alcohol
Formula: C₈H₆BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD09999512
CAS Number: 1026201-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR