2,4-Difluoro-5-methylbenzonitrile 97%

Warning

H31795-03 H31795-MD
AAH31795-03EA 131.36 USD
AAH31795-03 AAH31795-MD
2,4-Difluoro-5-methylbenzonitrile 97%
2,4-Difluoro-5-methylbenzonitrile
Formula: C₈H₅F₂N
MW: 153.13 g/mol
Melting Pt: 43…50 °C
Storage Temperature: Ambient
MDL Number: MFCD09835341
CAS Number: 329314-68-3
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR