8-Hydroxyjulolidine-9-carboxaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
9-Formyl-8-hydroxyjulolidine
H31782-03
H31782-06
AAH31782-06EA
201.99
USD
AAH31782-06
AAH31782-03
8-Hydroxyjulolidine-9-carboxaldehyde 97%
8-Hydroxyjulolidine-9-carboxaldehyde
Formula:
C₁₃H₁₅NO₂ MW: 217.27 g/mol Melting Pt: 72…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00192477 CAS Number: 63149-33-7 EINECS: 263-966-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...