3-Bromo-5-(trifluoromethoxy)benzonitrile 97%, JRD

Danger

H31778-03 H31778-06
AAH31778-03EA 99.56 USD
AAH31778-03 AAH31778-06
3-Bromo-5-(trifluoromethoxy)benzonitrile 97%, JRD
3-Bromo-5-(trifluoromethoxy)benzonitrile
Formula: C₈H₃BrF₃NO
MW: 266.01 g/mol
Melting Pt: 35…38 °C
Storage Temperature: Ambient
MDL Number: MFCD08458074
CAS Number: 914635-52-2
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR