2-Aminobenzenesulfonic acid 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Aniline-2-sulphonic acid
, Orthanilic acid
H31763-22
AAH31763-22EA
62.29
USD
AAH31763-22
2-Aminobenzenesulfonic acid 95%
2-Aminobenzenesulfonic acid
Formula:
C₆H₇NO₃S MW: 173.19 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007705 CAS Number: 88-21-1 EINECS: 201-810-9 UN: 2585 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...