2-Chloro-3-fluorobenzoic acid 97%

Warning

H31761-03 H31761-06
AAH31761-06EA 228.69 USD
AAH31761-06 AAH31761-03
2-Chloro-3-fluorobenzoic acid 97%
2-Chloro-3-fluorobenzoic acid
Formula: C₇H₄ClFO₂
Melting Pt: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00973903
CAS Number: 102940-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR