Methyl-3-cyano-5-fluorobenzoate 96%

Warning

Synonyms: Methyl 3-cyano-5-fluorobenzoate

H31759-03 H31759-MD
AAH31759-03EA 166.5 USD
AAH31759-03 AAH31759-MD
Methyl-3-cyano-5-fluorobenzoate 96%
Methyl 3-cyano-5-fluorobenzoate
Formula: C₉H₆FNO₂
MW: 179.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08688582
CAS Number: 886732-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR