5-Bromo-2-(trifluoromethoxy)benzaldehyde 98%

Supplier: Thermo Scientific Chemicals

Warning

H31751-03 H31751-09
AAH31751-09EA 333 USD
AAH31751-09 AAH31751-03
5-Bromo-2-(trifluoromethoxy)benzaldehyde 98%
5-Bromo-2-(trifluoromethoxy)benzaldehyde
Formula: C₈H₄BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD03701029
CAS Number: 923281-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR