2-Chloro-6-(trifluoromethyl)benzonitrile 97%, JRD

Warning

H31731-06 H31731-03
AAH31731-03EA 93.68 USD
AAH31731-03 AAH31731-06
2-Chloro-6-(trifluoromethyl)benzonitrile 97%, JRD
2-Chloro-6-(trifluoromethyl)benzonitrile
Formula: C₈H₃ClF₃N
MW: 205.57 g/mol
Melting Pt: 52…54 °C
Storage Temperature: Ambient
MDL Number: MFCD03412191
CAS Number: 129604-28-0
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR