4-Methoxy-3-(trifluoromethyl)phenylacetonitrile 97%, JRD

Supplier: Alfa Aesar
Warning

H31730-03 H31730-MD
AAH31730-03EA 132.88 USD
AAH31730-03 AAH31730-MD
4-Methoxy-3-(trifluoromethyl)phenylacetonitrile 97%, JRD
4-Methoxy-3-(trifluoromethyl)phenylacetonitrile
Formula: C₁₀H₈F₃NO
Melting Pt: 50…53 °C
Storage Temperature: Ambient
MDL Number: MFCD09832301
CAS Number: 1000555-32-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR