4-Bromo-2-(trifluoromethoxy)phenol 98%

Supplier: Thermo Scientific Chemicals
Warning

H31726-03 H31726-06
AAH31726-06EA 386.99 USD
AAH31726-06 AAH31726-03
4-Bromo-2-(trifluoromethoxy)phenol 98%
4-Bromo-2-(trifluoromethoxy)phenol
Formula: C₇H₄BrF₃O₂
Melting Pt: 51…55 °C
Storage Temperature: Ambient
MDL Number: MFCD09037798
CAS Number: 690264-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR