4-Ethoxy-2,6-difluoroaniline 97%, JRD

Warning

H31722-MD
AAH31722-MDEA 128.76 USD
AAH31722-MD
4-Ethoxy-2,6-difluoroaniline 97%, JRD
4-Ethoxy-2,6-difluoroaniline
Formula: C₈H₉F₂NO
Storage Temperature: Ambient
MDL Number: MFCD09832394
CAS Number: 1017779-79-1
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR