2,3-Dibromoisonicotinic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,3-Dibromopyridine-4-carboxylic acid

H31708-03
AAH31708-03EA 318.99 USD
AAH31708-03
2,3-Dibromoisonicotinic acid 97%
2,3-Dibromoisonicotinic acid
Formula: C₆H₃Br₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD09997708
CAS Number: 1020056-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR