2-(Difluoromethoxy)-5-fluorobenzonitrile 95%, JRD

Warning

H31695-03 H31695-MD
AAH31695-03EA 174.27 USD
AAH31695-03 AAH31695-MD
2-(Difluoromethoxy)-5-fluorobenzonitrile 95%, JRD
2-(Difluoromethoxy)-5-fluorobenzonitrile
Formula: C₈H₄F₃NO
Storage Temperature: Ambient
MDL Number: MFCD09832329
CAS Number: 1017778-48-1
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR