2-Chloro-1-methylimidazole-5-boronic acid pinacol ester 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Chloro-1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazole
H31692-MD
AAH31692-MDEA
280.99
USD
AAH31692-MD
2-Chloro-1-methylimidazole-5-boronic acid pinacol ester 95%
2-Chloro-1-methylimidazole-5-boronic acid pinacol ester
Formula:
C₁₀H₁₆BClN₂O₂ MW: 242.52 g/mol Melting Pt: 131…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD11100959 CAS Number: 1315281-29-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...