4'-Hydroxy-3'-(trifluoromethyl)acetophenone 95%

Supplier: Thermo Scientific Chemicals

Warning

H31691-MD H31691-03
AAH31691-03EA 132 USD
AAH31691-03 AAH31691-MD
4'-Hydroxy-3'-(trifluoromethyl)acetophenone 95%
4'-Hydroxy-3'-(trifluoromethyl)acetophenone
Formula: C₉H₇F₃O₂
Melting Pt: 174…177 °C
Storage Temperature: Ambient
MDL Number: MFCD01091005
CAS Number: 149105-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR