5-Nitro-3-picoline 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Methyl-5-nitropyridine

H31683-06 H31683-03
AAH31683-03EA 97.51 USD
AAH31683-03 AAH31683-06
5-Nitro-3-picoline 97%
5-Nitro-3-picoline
Formula: C₆H₆N₂O₂
MW: 138.13
Melting Pt: 94…98 °C
Storage Temperature: Ambient
MDL Number: MFCD04114214
CAS Number: 6960-20-9
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR