3-Chloro-4-ethoxy-5-fluorobenzylamine 97%, JRD

Danger

H31681-03 H31681-MD
AAH31681-03EA 247.53 USD
AAH31681-03 AAH31681-MD
3-Chloro-4-ethoxy-5-fluorobenzylamine 97%, JRD
3-Chloro-4-ethoxy-5-fluorobenzylamine
Formula: C₉H₁₁ClFNO
Storage Temperature: Ambient
MDL Number: MFCD09832346
CAS Number: 1017779-02-0
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR