Nujol (light oil)

Warning

Synonyms: Light liquid paraffinMineral oilVaseline oilParaffin, liquid

H31661-0F H31661-AD H31661-AP
AAH31661-0FEA 136.53 USD
AAH31661-0F AAH31661-AP AAH31661-AD
Nujol (light oil)
Nujol
Boiling Pt: >300 °C
Melting Pt: ca …20 °C
Density: 0.838
Flash Pt: 220 °C(428 °F)
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8012-95-1
EINECS: 232-384-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR