2,4-Difluoro-3-methylbenzoyl chloride 98%

Danger

H31658-03 H31658-MD
AAH31658-03EA 184.12 USD
AAH31658-03 AAH31658-MD
2,4-Difluoro-3-methylbenzoyl chloride 98%
2,4-Difluoro-3-methylbenzoyl chloride
Formula: C₈H₅ClF₂O
MW: 190.58 g/mol
Melting Pt: 35…38 °C
Storage Temperature: Ambient
MDL Number: MFCD07777157
CAS Number: 112857-70-2
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR