2-Methoxy-4,6-bis(trifluoromethyl)benzonitrile 97%, JRD

Danger

H31652-03 H31652-MD
AAH31652-03EA 197.58 USD
AAH31652-03 AAH31652-MD
2-Methoxy-4,6-bis(trifluoromethyl)benzonitrile 97%, JRD
2-Methoxy-4,6-bis(trifluoromethyl)benzonitrile
Formula: C₁₀H₅F₆NO
Melting Pt: 46…48 °C
Storage Temperature: Ambient
MDL Number: MFCD11519321
CAS Number: 1092460-75-7
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR