2-Fluoro-4-methoxybenzoyl chloride 95%, JRD

Danger

H31644-06 H31644-03
AAH31644-06EA 613.91 USD
AAH31644-06 AAH31644-03
2-Fluoro-4-methoxybenzoyl chloride 95%, JRD
2-Fluoro-4-methoxybenzoyl chloride
Formula: C₈H₆ClFO₂
Melting Pt: 40…42 °C
Storage Temperature: Ambient
MDL Number: MFCD08146649
CAS Number: 321-24-4
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR