3,4'-Bis(trifluoromethyl)benzhydrol 97%

Warning

H31629-03 H31629-06
AAH31629-06EA 205.35 USD
AAH31629-06 AAH31629-03
3,4'-Bis(trifluoromethyl)benzhydrol 97%
3,4'-Bis(trifluoromethyl)benzhydrol
Formula: C₁₅H₁₀F₆O
Storage Temperature: Ambient
MDL Number: MFCD00045091
CAS Number: 203915-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR