1-Fluoro-3-iodo-5-nitrobenzene 97%

Warning

H31612-03
AAH31612-03EA 72.25 USD
AAH31612-03
1-Fluoro-3-iodo-5-nitrobenzene 97%
1-Fluoro-3-iodo-5-nitrobenzene
Formula: C₆H₃FINO₂
MW: 267 g/mol
Melting Pt: 77…78.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00007219
CAS Number: 3819-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR