3-Phenyl-5-isoxazolecarboxylic acid 97%

Warning

Synonyms: 3-Phenylisoxazole-5-carboxylic acid

H31606-03
AAH31606-03EA 69.66 USD
AAH31606-03
3-Phenyl-5-isoxazolecarboxylic acid 97%
3-Phenyl-5-isoxazolecarboxylic acid
Formula: C₁₀H₇NO₃
MW: 189.17
Melting Pt: 180…182 °C
Storage Temperature: Ambient
MDL Number: MFCD01936010
CAS Number: 14442-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR