1-(Chloromethyl)-2-(trifluoromethoxy)benzene 98%

Danger

Synonyms: 2-(Trifluoromethoxy)benzyl chloride

H31603-03 H31603-09
AAH31603-09EA 245.31 USD
AAH31603-09 AAH31603-03
1-(Chloromethyl)-2-(trifluoromethoxy)benzene 98%
1-(Chloromethyl)-2-(trifluoromethoxy)benzene
Formula: C₈H₆ClF₃O
MW: 210.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00221456
CAS Number: 116827-40-8
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR