1-[2-(Trifluoromethyl)phenyl]propanol 98%

H31591-06 H31591-03
AAH31591-06EA 245.31 USD
AAH31591-06 AAH31591-03
1-[2-(Trifluoromethyl)phenyl]propanol 98%
1-[2-(Trifluoromethyl)phenyl]propanol
Formula: C₁₀H₁₁F₃O
MW: 204.19
Storage Temperature: Ambient
MDL Number: MFCD08062381
CAS Number: 878572-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR