3-Ethoxy-2,6-difluorobenzylamine 97%, JRD

Danger

H31587-MD H31587-03
AAH31587-03EA 310.8 USD
AAH31587-03 AAH31587-MD
3-Ethoxy-2,6-difluorobenzylamine 97%, JRD
3-Ethoxy-2,6-difluorobenzylamine
Formula: C₉H₁₁F₂NO
Storage Temperature: Ambient
MDL Number: MFCD11519371
CAS Number: 1092461-29-4
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR