4-(4-Bromophenyl)imidazole 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H31581-06
AAH31581-06EA
89.59
USD
AAH31581-06
4-(4-Bromophenyl)imidazole 97%
4-(4-Bromophenyl)imidazole
Formula:
C₉H₇BrN₂ MW: 223.07 g/mol Melting Pt: 138…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD03312935 CAS Number: 13569-96-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...