5-Amino-3-(4-methoxyphenyl)-1H-pyrazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Amino-3-(4-methoxyphenyl)pyrazole3-(4-Methoxyphenyl)-1H-pyrazol-5-amine

H31580-03 H31580-06
AAH31580-03EA 50.99 USD
AAH31580-03 AAH31580-06
5-Amino-3-(4-methoxyphenyl)-1H-pyrazole 97%
5-Amino-3-(4-methoxyphenyl)-1H-pyrazole
Formula: C₁₀H₁₁N₃O
MW: 189.22 g/mol
Melting Pt: 141…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00462192
CAS Number: 19541-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR