4,5-Difluoro-2-methoxyaniline 97%, JRD

Warning

H31575-06 H31575-03
AAH31575-06EA 588.65 USD
AAH31575-06 AAH31575-03
4,5-Difluoro-2-methoxyaniline 97%, JRD
4,5-Difluoro-2-methoxyaniline
Formula: C₇H₇F₂NO
MW: 159.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832387
CAS Number: 1017779-71-3
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR