Hydrogen chloride ∼0.5 M in methanol

Supplier: Thermo Scientific Chemicals

Danger

H31570-AE H31570-AP
AAH31570-APEA 157.87 USD
AAH31570-AP AAH31570-AE
Hydrogen chloride ∼0.5 M in methanol
Hydrogen chloride
Formula: HCl
MW: 36.46 g/mol
Density: 0.8
Flash Pt: 12 °C(54 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 3286
ADR: "3",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR