2-Nitro-4-(trifluoromethyl)benzyl alcohol 97+%
|
Warning
|
Synonyms:
(2-Nitro-4-(trifluoromethyl)phenyl)methanol
H31559-03
H31559-06
AAH31559-03EA
89.73
USD
AAH31559-03
AAH31559-06
2-Nitro-4-(trifluoromethyl)benzyl alcohol 97+%
2-Nitro-4-(trifluoromethyl)benzyl alcohol
Formula:
C₈H₆F₃NO₃ MW: 221.13 g/mol Melting Pt: 63…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095418 CAS Number: 133605-27-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...