2,5-Difluorobenzenesulfonamide 98%

Warning

H31556-06 H31556-03
AAH31556-03EA 98.93 USD
AAH31556-03 AAH31556-06
2,5-Difluorobenzenesulfonamide 98%
2,5-Difluorobenzenesulphonamide
Formula: C₆H₅F₂NO₂S
MW: 193.17 g/mol
Melting Pt: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00798115
CAS Number: 120022-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR