(2-Methoxy-4-(trifluoromethyl)phenyl)acetic acid 97%, JRD

Warning

H31551-03 H31551-MD
AAH31551-03EA 213.12 USD
AAH31551-03 AAH31551-MD
(2-Methoxy-4-(trifluoromethyl)phenyl)acetic acid 97%, JRD
(2-Methoxy-4-(trifluoromethyl)phenyl)acetic acid
Formula: C₁₀H₉F₃O₃
MW: 234.17 g/mol
Melting Pt: 99.5…102 °C
Storage Temperature: Ambient
MDL Number: MFCD09832353
CAS Number: 1017779-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR