3,4-Difluoro-2-methylbenzonitrile 98%

Supplier: Thermo Scientific Chemicals

Danger

H31548-06 H31548-03
AAH31548-03EA 113.88 USD
AAH31548-03 AAH31548-06
3,4-Difluoro-2-methylbenzonitrile 98%
3,4-Difluoro-2-methylbenzonitrile
Formula: C₈H₅F₂N
MW: 153.13 g/mol
Melting Pt: 39…44 °C
Storage Temperature: Ambient
MDL Number: MFCD07777154
CAS Number: 847502-83-4
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR