Ethyl-2-chloro-4-oxazolecarboxylate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 2-chlorooxazole-4-carboxylate

H31521-MD H31521-03
AAH31521-MDEA 84.27 USD
AAH31521-MD AAH31521-03
Ethyl-2-chloro-4-oxazolecarboxylate 95%
Ethyl 2-chloro-4-oxazolecarboxylate
Formula: C₆H₆ClNO₃
MW: 175.57 g/mol
Melting Pt: 97…98 °C
Storage Temperature: Ambient
MDL Number: MFCD06660120
CAS Number: 460081-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR