4-(Trifluoromethylthio)benzylamine 95%

Supplier: Thermo Scientific Chemicals
Danger

H31515-03 H31515-06
AAH31515-03EA 132.99 USD
AAH31515-03 AAH31515-06
4-(Trifluoromethylthio)benzylamine 95%
4-(Trifluoromethylthio)benzylamine
Formula: C₈H₈F₃NS
Boiling Pt: 224…225 °C
Density: 1.310
Flash Pt: 80 °C(176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00190129
CAS Number: 128273-56-3
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR