4-Methoxy-2-(trifluoromethyl)benzonitrile 97%, JRD

Danger

H31513-06 H31513-03
AAH31513-03EA 76.36 USD
AAH31513-03 AAH31513-06
4-Methoxy-2-(trifluoromethyl)benzonitrile 97%, JRD
4-Methoxy-2-(trifluoromethyl)benzonitrile
Formula: C₉H₆F₃NO
Melting Pt: 63…66 °C
Storage Temperature: Ambient
MDL Number: MFCD06797342
CAS Number: 875664-48-5
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR