4-(4-Fluorophenyl)imidazole 97%

Supplier: Thermo Scientific Chemicals
Danger

H31503-06
AAH31503-06EA 88.79 USD
AAH31503-06
4-(4-Fluorophenyl)imidazole 97%
4-(4-Fluorophenyl)imidazole
Formula: C₉H₇FN₂
MW: 162.17 g/mol
Melting Pt: 127…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00297041
CAS Number: 65020-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR