2-(4-Phenoxyphenyl)ethylboronic acid diethanolamine ester 97%

Warning

H31401-03
AAH31401-03EA 60.38 USD
AAH31401-03
2-(4-Phenoxyphenyl)ethylboronic acid diethanolamine ester 97%
2-(4-Phenoxyphenyl)ethylboronic acid diethanolamine ester
Formula: C₁₈H₂₂BNO₃
Melting Pt: 164…167 °C
Storage Temperature: Ambient
MDL Number: MFCD12546199
CAS Number: 608534-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR