2-Fluoro-4-methylbenzoyl chloride 97%
|
Danger
|
H31292-06
H31292-03
AAH31292-03EA
105.67
USD
AAH31292-03
AAH31292-06
2-Fluoro-4-methylbenzoyl chloride 97%
2-Fluoro-4-methylbenzoyl chloride
Formula:
C₈H₆ClFO MW: 172.59 g/mol Melting Pt: 43…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115879 CAS Number: 59189-98-9 UN: 3261 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...