2-Bromomalondialdehyde 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Bromomalonaldehyde
, 2-Bromomalonaldehyde
H31030-06
H31030-03
H31030-18
AAH31030-03EA
41.52
USD
AAH31030-03
AAH31030-18
AAH31030-06
2-Bromomalondialdehyde 97%
2-Bromomalondialdehyde
Formula:
C₃H₃BrO₂ MW: 150.96 g/mol Melting Pt: ca 145 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00459999 CAS Number: 2065-75-0 EINECS: 430-470-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...