2-Iodoisonicotinic acid 95%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 2-Iodopyridine-4-carboxylic acid

H31002-03 H31002-MD
AAH31002-MDEA 58.05 USD
AAH31002-MD AAH31002-03
2-Iodoisonicotinic acid 95%
2-Iodoisonicotinic acid
Formula: C₆H₄INO₂
Melting Pt: ca 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD06800803
CAS Number: 58481-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR