5-Fluoro-2-methoxypyridine 97%

Warning

H30991-14 H30991-06 H30991-03
AAH30991-06EA 165.39 USD
AAH30991-06 AAH30991-14 AAH30991-03
5-Fluoro-2-methoxypyridine 97%
5-Fluoro-2-methoxypyridine
Formula: C₆H₆FNO
Storage Temperature: Refrigerator
MDL Number: MFCD03095252
CAS Number: 51173-04-7
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR